siRNA Details
Gene Symbol: MSANTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042690.1 | NP_001036155.1 | 3 | 1319 | ||
XM_011513465.2 | XP_011511767.1 | 6 | 2581 | |||
XM_011513466.2 | XP_011511768.1 | 4 | 4846 | |||
XM_011513467.2 | XP_011511769.1 | 2 | 6355 | |||
XM_017008169.1 | XP_016863658.1 | 4 | 4024 | |||
GenBank mRNA | BC043530.1 | - | 6 | 1940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
