siRNA Details
Gene Symbol: MSANTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042690.1 | NP_001036155.1 | 2 | 563 | ||
XM_011513465.2 | XP_011511767.1 | 5 | 1825 | |||
XM_011513466.2 | XP_011511768.1 | 3 | 4090 | |||
XM_017008169.1 | XP_016863658.1 | 3 | 3268 | |||
GenBank mRNA | AK126094.1 | - | 2 | 563 | ||
BC043530.1 | - | 5 | 1184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
