siRNA Details
Gene Symbol: SIGLEC17P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047529.2 | 3 | 530 | |||
NR_047530.2 | 3 | 530 | ||||
NR_047531.2 | 3 | 530 | ||||
GenBank mRNA | AF150143.1 | - | 3 | 460 | ||
AF161341.1 | - | 3 | 460 | |||
AK301336.1 | - | 3 | 460 | |||
BC041072.1 | - | 3 | 530 | |||
BC050999.1 | - | 3 | 530 | |||
DC340819.1 | - | 3 | 460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
