siRNA Details
Gene Symbol: ARSI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012301.2 | NP_001012301.1 | 2 | 1601 | ||
GenBank mRNA | AB448735.1 | - | 2 | 1592 | ||
AK122641.1 | - | 2 | 745 | |||
AY875937.1 | - | 2 | 1012 | |||
BC129995.1 | - | 2 | 1069 | 40127932 | ||
BC129996.1 | - | 2 | 1069 | 40127934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
