siRNA Details
Gene Symbol: OTOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277269.1 | NP_001264198.1 | 46 | 7801 | ||
NM_001292063.1 | NP_001278992.1 | 47 | 7765 | |||
GenBank mRNA | AK128214.1 | - | 20 | 4677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
