siRNA Details
Gene Symbol: FCHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146032.1 | NP_001139504.1 | 6 | 688 | ||
NM_138782.2 | NP_620137.2 | 6 | 688 | |||
XM_017009017.1 | XP_016864506.1 | 6 | 682 | |||
XM_017009018.1 | XP_016864507.1 | 6 | 682 | |||
XM_017009019.1 | XP_016864508.1 | 6 | 682 | |||
XM_017009020.1 | XP_016864509.1 | 6 | 682 | |||
XM_017009021.1 | XP_016864510.1 | 6 | 682 | |||
XM_017009022.1 | XP_016864511.1 | 6 | 682 | |||
XM_017009023.1 | XP_016864512.1 | 6 | 682 | |||
GenBank mRNA | AK291782.1 | - | 6 | 618 | ||
AK295141.1 | - | 6 | 598 | |||
AL831971.1 | - | 6 | 597 | |||
AL833313.1 | - | 4 | 389 | |||
BC014311.1 | - | 6 | 602 | |||
BC137070.1 | - | 6 | 688 | 9021067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
