siRNA Details
Gene Symbol: SPATA31D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145197.1 | NP_001138669.1 | 4 | 2265 | ||
GenBank mRNA | AK125847.1 | - | 4 | 2210 | ||
AK308327.1 | - | 4 | 2206 |
Gene Symbol: SPATA31D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207416.2 | NP_997299.2 | 4 | 2265 | ||
GenBank mRNA | AK126070.1 | - | 1 | 968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
