siRNA Details
Gene Symbol: SBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024401.2 | NP_001019572.1 | 2 | 946 | ||
XM_005255315.3 | XP_005255372.1 | 2 | 980 | |||
GenBank mRNA | AY874862.2 | - | 2 | 946 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
