siRNA Details
Gene Symbol: PIRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101387.1 | NP_001094857.1 | 2 | 334 | ||
GenBank mRNA | AK021549.1 | - | 2 | 315 | ||
AK299808.1 | - | 2 | 320 | |||
DC332889.1 | - | 2 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
