siRNA Details
Gene Symbol: FAM168B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009993.3 | NP_001009993.2 | 3 | 388 | ||
NM_001321743.1 | NP_001308672.1 | 5 | 837 | |||
NM_001321744.1 | NP_001308673.1 | 4 | 543 | |||
NM_001321745.1 | NP_001308674.1 | 3 | 291 | |||
NM_001321746.1 | NP_001308675.1 | 3 | 270 | |||
NM_001321747.1 | NP_001308676.1 | 3 | 211 | |||
NM_001321748.1 | NP_001308677.1 | 3 | 160 | |||
XM_011510594.2 | XP_011508896.1 | 2 | 203 | |||
XM_017003327.1 | XP_016858816.1 | 5 | 880 | |||
XM_017003328.1 | XP_016858817.1 | 4 | 494 | |||
GenBank mRNA | AY253283.1 | - | 2 | 133 | ||
BC066347.1 | - | 3 | 347 | 5312158 | ||
BC110642.1 | - | 3 | 238 | 6144610 | ||
CN418443.1 | - | 3 | 388 | |||
DB453209.1 | - | 3 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
