siRNA Details
Gene Symbol: FAM168B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009993.3 | NP_001009993.2 | 2 | 314 | ||
NM_001321743.1 | NP_001308672.1 | 4 | 763 | |||
NM_001321744.1 | NP_001308673.1 | 3 | 469 | |||
NM_001321745.1 | NP_001308674.1 | 2 | 217 | |||
NM_001321746.1 | NP_001308675.1 | 2 | 196 | |||
NM_001321747.1 | NP_001308676.1 | 2 | 137 | |||
NM_001321748.1 | NP_001308677.1 | 2 | 86 | |||
XM_017003327.1 | XP_016858816.1 | 4 | 806 | |||
XM_017003328.1 | XP_016858817.1 | 2 | 322 | |||
GenBank mRNA | AY253283.1 | - | 1 | 59 | ||
BC066347.1 | - | 2 | 273 | 5312158 | ||
BC110642.1 | - | 2 | 164 | 6144610 | ||
CN418443.1 | - | 2 | 314 | |||
DB453209.1 | - | 2 | 285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
