siRNA Details
Gene Symbol: TBC1D3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027486.1 | 3, 4 | 264 | |||
GenBank mRNA | AK301746.1 | - | 3, 4 | 265 | ||
BC058890.1 | - | 3, 4 | 236 | |||
EG327356.1 | - | 3, 4 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
