siRNA Details
Gene Symbol: FAM166B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099951.3 | NP_001093421.1 | 2 | 196 | ||
NM_001164310.2 | NP_001157782.1 | 2 | 196 | |||
NM_001287238.1 | NP_001274167.1 | 2 | 196 | |||
NM_001287239.1 | NP_001274168.1 | 2 | 196 | |||
XM_011518028.2 | XP_011516330.1 | 2 | 335 | |||
GenBank mRNA | BC129999.1 | - | 2 | 196 | 40127800 | |
BC146928.1 | - | 2 | 196 | 9056856 | ||
BC146933.1 | - | 2 | 196 | 9056861 | ||
BC171840.1 | - | 2 | 196 | 9054494 | ||
BC171851.1 | - | 2 | 196 | 9054505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
