siRNA Details
Gene Symbol: TMEM91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042595.2 | NP_001036060.1 | 3 | 351 | ||
NM_001098821.1 | NP_001092291.1 | 3 | 613 | |||
NM_001098822.1 | NP_001092292.1 | 3 | 613 | |||
NM_001098823.1 | NP_001092293.1 | 3 | 613 | |||
NM_001098824.1 | NP_001092294.1 | 3 | 613 | |||
NM_001098825.1 | NP_001092295.1 | 3 | 421 | |||
GenBank mRNA | AK130820.1 | - | 3 | 922 | ||
AL833095.1 | - | 3 | 583 | |||
BC063705.1 | - | 3 | 323 | 4804092 | ||
BF684543.1 | - | 3 | 413 | |||
BG697182.1 | - | 3 | 351 | |||
BM726200.1 | - | 3 | 388 | |||
BM931402.1 | - | 3 | 568 | |||
BX090160.1 | - | 2 | 296 | |||
R67551.1 | - | 2 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
