siRNA Details
Gene Symbol: HACD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010915.4 | NP_001010915.2 | 7 | 807 | ||
NM_001321883.1 | NP_001308812.1 | 7 | 744 | |||
NM_001321903.1 | NP_001308832.1 | 9 | 960 | |||
XM_011517876.2 | XP_011516178.1 | 8 | 835 | |||
XM_017014713.1 | XP_016870202.1 | 9 | 1150 | |||
XM_017014714.1 | XP_016870203.1 | 8 | 897 | |||
GenBank mRNA | BC114215.1 | - | 7 | 741 | 5174279 | |
BX648779.1 | - | 7 | 769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
