siRNA Details
Gene Symbol: HACD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010915.4 | NP_001010915.2 | 7 | 1795 | ||
NM_001321883.1 | NP_001308812.1 | 7 | 1732 | |||
NM_001321903.1 | NP_001308832.1 | 9 | 1948 | |||
XM_011517876.2 | XP_011516178.1 | 8 | 1823 | |||
XM_017014713.1 | XP_016870202.1 | 9 | 2138 | |||
XM_017014714.1 | XP_016870203.1 | 8 | 1885 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
