siRNA Details
Gene Symbol: SMCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077657.2 | NP_001071125.1 | 3 | 398 | ||
NM_001320473.1 | NP_001307402.1 | 3 | 454 | |||
GenBank mRNA | AK123917.1 | - | 3 | 398 | ||
AK291522.1 | - | 3 | 408 | |||
AL833140.1 | - | 3 | 379 | |||
AL833443.1 | - | 3 | 434 | |||
BC118556.1 | - | 3 | 360 | 40023903 | ||
BC118636.1 | - | 3 | 360 | 40023902 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
