siRNA Details
Gene Symbol: C2orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080474.1 | NP_001073943.1 | 3 | 1251 | ||
XM_005264302.3 | XP_005264359.1 | 3 | 1196 | |||
GenBank mRNA | AK125975.1 | - | 3 | 1252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
