siRNA Details
Gene Symbol: ATP13A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198505.2 | NP_940907.2 | 24 | 2737 | ||
XM_011512770.1 | XP_011511072.1 | 24 | 2836 | |||
XM_017006304.1 | XP_016861793.1 | 20 | 2050 | |||
XM_017006305.1 | XP_016861794.1 | 11 | 1153 | |||
GenBank mRNA | AY823163.1 | - | 24 | 2737 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
