siRNA Details
Gene Symbol: ATP13A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198505.2 | NP_940907.2 | 28, 29 | 3323 | ||
XM_017006304.1 | XP_016861793.1 | 24, 25 | 2636 | |||
XM_017006305.1 | XP_016861794.1 | 15, 16 | 1739 | |||
GenBank mRNA | AY358667.1 | - | 2, 3 | 132 | ||
AY823163.1 | - | 28, 29 | 3323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
