siRNA Details
Gene Symbol: C5orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145132.1 | NP_001138604.1 | 2 | 360 | ||
XM_011534416.1 | XP_011532718.1 | 3 | 386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
