siRNA Details
Gene Symbol: C5orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145132.1 | NP_001138604.1 | 3 | 430 | ||
XM_011534416.1 | XP_011532718.1 | 4 | 456 | |||
XM_017008909.1 | XP_016864398.1 | 2 | 495 | |||
GenBank mRNA | BG721329.1 | - | 3 | 253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
