siRNA Details
Gene Symbol: SMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003652.3 | NP_001003652.1 | 11 | 9916 | ||
NM_001135937.2 | NP_001129409.1 | 10 | 9826 | |||
NM_005901.5 | NP_005892.1 | 11 | 9808 | |||
XM_005258259.3 | XP_005258316.1 | 12 | 10049 | |||
XM_006722451.3 | XP_006722514.1 | 12 | 10061 | |||
XM_011525984.2 | XP_011524286.1 | 11 | 9405 | |||
XM_011525985.2 | XP_011524287.1 | 13 | 10108 | |||
XM_017025745.1 | XP_016881234.1 | 13 | 10147 | |||
XM_017025746.1 | XP_016881235.1 | 11 | 9959 | |||
XM_017025747.1 | XP_016881236.1 | 12 | 10034 | |||
XM_017025748.1 | XP_016881237.1 | 13 | 10120 | |||
XM_017025750.1 | XP_016881239.1 | 12 | 10009 | |||
GenBank mRNA | AY134745.1 | - | 1 | 445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
