siRNA Details
Gene Symbol: SLC9A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284351.2 | NP_001271280.1 | 17 | 3100 | ||
NM_004174.3 | NP_004165.2 | 17 | 3127 |
Gene Symbol: PP7080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024158.1 | 1 | 97 | |||
GenBank mRNA | AF370427.1 | - | 1 | 195 | ||
BC072677.1 | - | 1 | 93 | |||
BC086861.1 | - | 1 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
