siRNA Details
Gene Symbol: PP7080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024158.1 | 2 | 1704 | |||
GenBank mRNA | AF370427.1 | - | 2 | 1805 | ||
BC086861.1 | - | 2 | 1685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
