siRNA Details
Gene Symbol: LRRC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291006.1 | NP_001277935.1 | 5 | 709 | ||
NR_027006.1 | 5 | 1786 | ||||
NR_110991.1 | 4 | 1643 | ||||
XM_011530173.2 | XP_011528475.1 | 5 | 688 | |||
GenBank mRNA | AK128646.1 | - | 5 | 1765 | ||
BC140948.1 | - | 4 | 1641 | |||
BC140950.1 | - | 4 | 1641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
