siRNA Details
Gene Symbol: SNED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080437.1 | NP_001073906.1 | 32 | 5101 | ||
XM_011510931.2 | XP_011509233.1 | 33 | 5415 | |||
XM_011510932.2 | XP_011509234.1 | 32 | 5318 | |||
GenBank mRNA | AF439717.1 | - | 26 | 3944 | ||
AK074062.1 | - | 31 | 4709 | |||
AK074075.1 | - | 12 | 2428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
