siRNA Details
Gene Symbol: SNED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080437.1 | NP_001073906.1 | 22 | 3181 | ||
XM_011510931.2 | XP_011509233.1 | 23 | 3495 | |||
XM_011510932.2 | XP_011509234.1 | 23 | 3497 | |||
XM_011510933.2 | XP_011509235.1 | 23 | 3501 | |||
XM_011510934.2 | XP_011509236.1 | 23 | 3501 | |||
XM_017003773.1 | XP_016859262.1 | 23 | 3497 | |||
XM_017003774.1 | XP_016859263.1 | 22 | 3266 | |||
XM_017003775.1 | XP_016859264.1 | 23 | 3499 | |||
XM_017003776.1 | XP_016859265.1 | 22 | 3383 | |||
XM_017003777.1 | XP_016859266.1 | 21 | 3274 | |||
XM_017003778.1 | XP_016859267.1 | 22 | 3261 | |||
XM_017003779.1 | XP_016859268.1 | 23 | 3197 | |||
XM_017003780.1 | XP_016859269.1 | 22 | 3053 | |||
XM_017003781.1 | XP_016859270.1 | 22 | 3043 | |||
XM_017003782.1 | XP_016859271.1 | 18 | 2392 | |||
GenBank mRNA | AF439717.1 | - | 16 | 2084 | ||
AF439718.1 | - | 2 | 370 | |||
AK074062.1 | - | 21 | 2789 | |||
AK074075.1 | - | 4 | 691 | |||
BC027939.1 | - | 5 | 1019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
