siRNA Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013659.2 | NP_001013681.2 | 8 | 951 | ||
XM_005258927.3 | XP_005258984.1 | 8 | 1063 | |||
XM_006723213.3 | XP_006723276.1 | 7 | 973 | |||
GenBank mRNA | AK131417.1 | - | 8 | 951 | ||
AK303740.1 | - | 6 | 744 | |||
BX537524.1 | - | 8 | 1427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
