siRNA Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013659.2 | NP_001013681.2 | 8 | 2761 | ||
XM_005258927.3 | XP_005258984.1 | 8 | 2873 | |||
XM_006723213.3 | XP_006723276.1 | 7 | 2783 | |||
GenBank mRNA | BX537524.1 | - | 8 | 3236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
