siRNA Details
Gene Symbol: USP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015306.2 | NP_056121.2 | 3 | 771 | ||
XM_017000831.1 | XP_016856320.1 | 3 | 776 | |||
XM_017000832.1 | XP_016856321.1 | 3 | 773 | |||
XM_017000833.1 | XP_016856322.1 | 3 | 268 | |||
XM_017000834.1 | XP_016856323.1 | 3 | 268 | |||
XM_017000835.1 | XP_016856324.1 | 3 | 779 | |||
XM_017000836.1 | XP_016856325.1 | 3 | 779 | |||
XM_017000837.1 | XP_016856326.1 | 3 | 779 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
