siRNA Details
Gene Symbol: C15orf61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143936.1 | NP_001137408.1 | 1, 2 | 414 | ||
GenBank mRNA | AK294012.1 | - | 1, 2 | 349 | ||
AY726587.1 | - | 1, 2 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
