siRNA Details
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 23 | 2937 | ||
NM_173636.4 | NP_775907.4 | 23 | 2937 | |||
XM_005258809.2 | XP_005258866.1 | 23 | 2937 | |||
XM_011526837.1 | XP_011525139.1 | 23 | 2922 | |||
XM_011526838.1 | XP_011525140.1 | 23 | 2937 | |||
XM_011526839.1 | XP_011525141.1 | 21 | 2586 | |||
XM_011526840.2 | XP_011525142.1 | 16 | 1932 | |||
XM_011526841.2 | XP_011525143.1 | 14 | 1553 | |||
XM_011526842.1 | XP_011525144.1 | 13 | 1385 | |||
XM_011526843.1 | XP_011525145.1 | 6 | 712 | |||
XM_011526844.2 | XP_011525146.1 | 6 | 709 | |||
XM_017026665.1 | XP_016882154.1 | 23 | 2937 | |||
GenBank mRNA | AL133651.1 | - | 2 | 254 | ||
BC017261.2 | - | 5 | 646 | |||
BC058939.1 | - | 23 | 2881 | |||
KU178897.1 | - | 2 | 182 | |||
KU178898.1 | - | 2 | 182 | |||
KU178899.1 | - | 2 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
