siRNA Details
Gene Symbol: CARMIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138360.3 | NP_612369.3 | 8 | 723 | ||
XM_011537300.2 | XP_011535602.1 | 7 | 382 | |||
XM_017021751.1 | XP_016877240.1 | 4 | 283 | |||
GenBank mRNA | BU102125.1 | - | 4 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
