siRNA Details
Gene Symbol: CARMIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138360.3 | NP_612369.3 | 10 | 864 | ||
XM_011537300.2 | XP_011535602.1 | 9 | 523 | |||
XM_017021751.1 | XP_016877240.1 | 6 | 424 | |||
GenBank mRNA | BG913168.1 | - | 1 | 16 | ||
BG977591.1 | - | 1 | 77 | |||
DT932621.1 | - | 1 | 34 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
