siRNA Details
Gene Symbol: SAXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008226.1 | NP_001008227.1 | 2 | 231 | ||
GenBank mRNA | AK304339.1 | - | 3 | 343 | ||
AL833762.1 | - | 2 | 239 | |||
BC144647.1 | - | 2 | 453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
