siRNA Details
Gene Symbol: LRRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024652.4 | NP_078928.3 | 6, 7 | 1113 | ||
XM_011522012.2 | XP_011520314.1 | 8, 9 | 1387 | |||
XM_011522013.2 | XP_011520315.1 | 6, 7 | 1096 | |||
XM_011522014.2 | XP_011520316.1 | 6, 7 | 1311 | |||
XM_011522016.2 | XP_011520318.1 | 4, 5 | 588 | |||
XM_011522019.2 | XP_011520321.1 | 6, 7 | 1311 | |||
GenBank mRNA | AK130975.1 | - | 2, 3 | 328 | ||
BC144705.1 | - | 5, 6 | 784 | |||
BC171802.1 | - | 5, 6 | 784 | |||
DQ013130.1 | - | 5, 6 | 751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
