siRNA Details
Gene Symbol: LRRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024652.4 | NP_078928.3 | 7 | 1330 | ||
XM_011522012.2 | XP_011520314.1 | 9 | 1604 | |||
XM_011522013.2 | XP_011520315.1 | 7 | 1313 | |||
XM_011522014.2 | XP_011520316.1 | 7 | 1528 | |||
XM_011522015.2 | XP_011520317.1 | 2 | 2123 | |||
XM_011522016.2 | XP_011520318.1 | 5 | 805 | |||
XM_011522019.2 | XP_011520321.1 | 7 | 1528 | |||
GenBank mRNA | AK130975.1 | - | 3 | 545 | ||
BC144705.1 | - | 6 | 1001 | |||
BC171802.1 | - | 6 | 1001 | |||
DQ013130.1 | - | 6 | 968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
