siRNA Details
Gene Symbol: ZNF385C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242704.1 | NP_001229633.1 | 3 | 229 | ||
XM_006721751.3 | XP_006721814.1 | 3 | 334 | |||
XM_011524479.2 | XP_011522781.1 | 4 | 1030 | |||
XM_011524482.2 | XP_011522784.1 | 4 | 1030 | |||
XM_011524483.2 | XP_011522785.1 | 4 | 1031 | |||
XM_017024314.1 | XP_016879803.1 | 4 | 1033 | |||
XM_017024315.1 | XP_016879804.1 | 4 | 868 | |||
XM_017024316.1 | XP_016879805.1 | 4 | 1033 | |||
XM_017024317.1 | XP_016879806.1 | 4 | 1034 | |||
XM_017024318.1 | XP_016879807.1 | 4 | 1035 | |||
XM_017024319.1 | XP_016879808.1 | 4 | 1031 | |||
XM_017024320.1 | XP_016879809.1 | 3 | 295 | |||
XM_017024321.1 | XP_016879810.1 | 4 | 1036 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
