siRNA Details
Gene Symbol: SPANXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009613.2 | NP_001009613.1 | 1 | 125 | ||
XM_017029543.1 | XP_016885032.1 | 1 | 115 | |||
XM_017029544.1 | XP_016885033.1 | 1 | 115 | |||
GenBank mRNA | BC130503.1 | - | 1 | 125 | 40146534 | |
BC130505.1 | - | 1 | 125 | 40146536 | ||
DQ336126.1 | - | 1 | 28 | |||
DQ336127.1 | - | 1 | 28 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
