siRNA Details
Gene Symbol: KIAA1324L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142749.2 | NP_001136221.1 | 2 | 405 | ||
NM_001291990.1 | NP_001278919.1 | 2 | 163 | |||
NM_152748.3 | NP_689961.3 | 2 | 198 | |||
XM_006715894.3 | XP_006715957.1 | 2 | 105 | |||
XM_011515918.2 | XP_011514220.1 | 2 | 256 | |||
XM_017011843.1 | XP_016867332.1 | 2 | 401 | |||
XM_017011844.1 | XP_016867333.1 | 2 | 191 | |||
GenBank mRNA | AK296250.1 | - | 2 | 115 | ||
AK303619.1 | - | 2 | 165 | |||
BX346336.2 | - | 2 | 409 | |||
DA240191.1 | - | 2 | 193 | |||
DY654933.1 | - | 2 | 72 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
