siRNA Details
Gene Symbol: ARHGEF33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145451.2 | NP_001138923.2 | 5 | 561 | ||
GenBank mRNA | DV080125.1 | - | 3 | 276 | ||
DY654603.1 | - | 1 | 39 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
