siRNA Details
Gene Symbol: TMEM178B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195278.1 | NP_001182207.1 | 2 | 542 | ||
XM_011515705.2 | XP_011514007.1 | 2 | 1372 | |||
XM_017011636.1 | XP_016867125.1 | 2 | 1366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
