siRNA Details
Gene Symbol: CFAP126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013625.3 | NP_001013647.2 | 5 | 407 | ||
GenBank mRNA | DT215043.1 | - | 5 | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
