siRNA Details
Gene Symbol: NEMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142645.1 | NP_001136117.1 | 1, 2 | 192 | ||
NR_136298.1 | 1, 2 | 192 | ||||
XM_005246212.3 | XP_005246269.1 | 1, 2 | 346 | |||
XM_005246213.3 | XP_005246270.1 | 1, 2 | 192 | |||
XM_011510458.2 | XP_011508760.1 | 1, 2 | 172 | |||
GenBank mRNA | AK296987.1 | - | 1, 2 | 140 | ||
AK302425.1 | - | 1, 2 | 317 | |||
DA417772.1 | - | 1, 2 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
