siRNA Details
Gene Symbol: LOC728613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK307076.1 | - | 2, 3 | 234 | ||
AK308990.1 | - | 2, 3 | 197 | |||
BC053586.1 | - | 2, 3 | 185 | |||
DA383480.1 | - | 2, 3 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
