siRNA Details
Gene Symbol: SZT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015284.3 | NP_056099.3 | 22 | 3207 | ||
XM_005270686.3 | XP_005270743.1 | 24 | 3439 | |||
XM_006710501.3 | XP_006710564.1 | 23 | 3326 | |||
XM_011541106.2 | XP_011539408.1 | 23 | 3477 | |||
XM_011541107.2 | XP_011539409.1 | 20 | 2974 | |||
XM_017000819.1 | XP_016856308.1 | 24 | 3439 | |||
XM_017000820.1 | XP_016856309.1 | 23 | 3268 | |||
XM_017000821.1 | XP_016856310.1 | 20 | 2960 | |||
GenBank mRNA | AB007936.2 | - | 8 | 1059 | ||
AK131107.1 | - | 14 | 1955 | |||
BC151232.1 | - | 8 | 1059 | 8860124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
