siRNA Details
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244752.1 | NP_001231681.1 | 14 | 2826 | ||
XM_011517881.2 | XP_011516183.1 | 15 | 2893 | |||
XM_011517883.2 | XP_011516185.1 | 14 | 2836 | |||
XM_011517885.2 | XP_011516187.1 | 9 | 1981 | |||
XM_017014728.1 | XP_016870217.1 | 9 | 1978 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
