siRNA Details
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244752.1 | NP_001231681.1 | 15 | 2934 | ||
XM_011517881.2 | XP_011516183.1 | 16 | 3001 | |||
XM_011517883.2 | XP_011516185.1 | 15 | 2944 | |||
XM_011517885.2 | XP_011516187.1 | 10 | 2089 | |||
XM_017014728.1 | XP_016870217.1 | 10 | 2086 |
Gene Symbol: ANKRD18CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136286.1 | 17 | 3096 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
