siRNA Details
Gene Symbol: ZNF777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015694.2 | NP_056509.2 | 3 | 1092 | ||
XM_005249980.3 | XP_005250037.1 | 3 | 1078 | |||
XM_011516055.2 | XP_011514357.1 | 3 | 1102 | |||
GenBank mRNA | AB033111.1 | - | 3 | 1006 | ||
AK075032.1 | - | 2 | 189 | |||
BC023985.2 | - | 2 | 156 | 2820826 | ||
BC105968.1 | - | 2 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
